Memmert™ Potential-Free Contact (24 V / 2 A) with Socket to NAMUR NE 28 for Combined Error Message for IN110plus
Potential-free contact for combination error message (e.g. supply failure, sensor fault, fuse).
Additional Details:
Weight:
0.01200kg
Specification
For Use With (Equipment) | IN110plus Incubator |
---|---|
Type | Potential-free Contact |
Includes | Socket to NAMUR NE 28 |
Voltage | 24 V |
Amperage | 2 A |