Brady™ Aggressive Adhesive, Multipurpose Clear Polyester Labels
Label circuit boards, panels and equipment with the B-432 clear polyester label material, allowing the applied surface colors to transmit around the printed label text. Print almost anywhere with a Brady™ handheld label maker.
Additional Details:
Weight:
0.15400kg
Specification
Certifications/Compliance | UL Recognised;CSA Accepted Compliant with Directive 2011/65/EU and 2015/863 |
---|---|
AlternatePartNumber | M5-17-432-CL-BK |
Adhesive Type | Permanent Acrylic |
Color | Transparent,Clear |
Color | Transparent,Clear |
Finish | Gloss |
Format | Die-cut |
Material | Polyester |
Product Type | Labels |
Sterility | Non-sterile |
For Use With (Application) | Asset tracking;Barcode labelling;Component and equipment labelling;Data and telecommunications labelling;Electrical labelling;General identification;Inventory and inspection labelling;Warehouse marking |
For Use With (Equipment) | BMP51;BMP53;M511;M510 |
Quantity | 280 |
Temperature Range (English) | -40°F to 212°F |
Temperature Range (Metric) | -40°C to 100°C |
Thickness (English) | 0.004 in |
Thickness (Metric) | 0.102 mm |
Dimensions (LxW) | 25.40 mm (W) x 12.70 mm (H) |
Length (English) | 0.5 in. |
Width (English) | 1 in |
Width (Metric) | 25.40 mm |
Height (English) | 0.5 in |
Height (Metric) | 12.70 mm |