522.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Disposable | Single-use |
---|---|
Filter Type | Ultrafiltration |
For Use With (Equipment) | Centrifuge |
Material (Membrane) | Regenerated Cellulose |
MWCO | 5 kDa |
Sample Volume | 15 mL |
Sterility | Non-sterile |
Housing Material | Polypropylene |
Quantity | 48/Pk. |
Type | Centrifugal Concentrator |
Volume (Metric) | 15 mL |
Length (Metric) | 77 mm |