Fisherbrand™ White Polypropylene Caps
White polypropylene screw caps provides clear surface for writing on cap
Additional Details:
Additional Details: Weight: 0.31500kg
Product Code. 16144607
 N/A
N/A
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
  • Description and Specification
  • Alternative Products
Description and Specification
Specification
Color White
Quantity 100
Closure Color White
Neck Finish 58-400
Closure Included No
Product Type Lined Cap
Closure Liner PTFE/Polyethylene Foam
Closure Material Polypropylene
  • Foamed polyethylene liner provides tight seal and chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
  • PTFE faced foamed polyethylene liner offers chemical resistance excellent for environmental samples, pharmaceuticals and diagnostic reagents
  • Poly-vinyl liner provides an excellent moisture barrier good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Alternative Products