Fisherbrand™ White Polypropylene Caps
White polypropylene screw caps provides clear surface for writing on cap
Additional Details:
Weight:
0.94500kg
Specification
Color | White |
---|---|
Quantity | 144 |
Closure Color | White |
Neck Finish | 89-400 |
Closure Included | No |
Product Type | Lined Cap |
Closure Liner | PTFE/Polyethylene Foam |
Closure Material | Polypropylene |