Fisherbrand™ White Polypropylene Caps
White polypropylene screw caps provides clear surface for writing on cap
Additional Details:
Weight:
8.88300kg
Specification
Color | White |
---|---|
Quantity | 1300 |
Closure Color | White |
Neck Finish | 53-400 |
Closure Included | No |
Product Type | Lined Cap |
Closure Liner | Polyethylene Foam |
Closure Material | Polypropylene |