Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | ??product.searchResults.label.quantityAvail?? |
|
|||
---|---|---|---|---|---|---|---|---|---|---|
10309612
|
Waters Corporation
186003117
|
250 mm | 4.6 mm | 5 μm |
£1218.0
Each |
|
||||
13222960
|
Waters Corporation
186003020
|
30 mm | 2.1 mm | 3.5 μm |
£970.5
Each |
|
||||
10729006
|
Waters Corporation
186003021
|
50 mm | 2.1 mm | 3.5 μm |
£999.0
Each |
|
||||
10614596
|
Waters Corporation
186003022
|
100 mm | 2.1 mm | 3.5 μm |
£1074.0
Each |
|
||||
13262960
|
Waters Corporation
186003025
|
30 mm | 3 mm | 3.5 μm |
£970.5
Each |
|
||||
13272960
|
Waters Corporation
186003026
|
50 mm | 3 mm | 3.5 μm |
£999.0
Each |
|
||||
10015683
|
Waters Corporation
186003027
|
100 mm | 3 mm | 3.5 μm |
£1074.0
Each |
|
||||
10136503
|
Waters Corporation
186003028
|
150 mm | 3 mm | 3.5 μm |
£1149.0
Each |
|
||||
13292960
|
Waters Corporation
186003030
|
30 mm | 4.6 mm | 3.5 μm |
£970.5
Each |
|
||||
10614776
|
Waters Corporation
186003031
|
50 mm | 4.6 mm | 3.5 μm |
£999.0
Each |
|
||||
13212970
|
Waters Corporation
186003032
|
75 mm | 4.6 mm | 3.5 μm |
£1017.0
Each |
|
||||
11916938
|
Waters Corporation
186003033
|
100 mm | 4.6 mm | 3.5 μm |
£1074.0
Each |
|
||||
10779186
|
Waters Corporation
186003034
|
150 mm | 4.6 mm | 3.5 μm |
£1149.0
Each |
|
||||
13203020
|
Waters Corporation
186003107
|
20 mm | 2.1 mm | 5 μm |
£895.5
Each |
|
||||
13213020
|
Waters Corporation
186003108
|
50 mm | 2.1 mm | 5 μm |
£964.5
Each |
|
||||
13253020
|
Waters Corporation
186003113
|
50 mm | 4.6 mm | 5 μm |
£964.5
Each |
|
||||
13263020
|
Waters Corporation
186003114
|
75 mm | 4.6 mm | 5 μm |
£999.0
Each |
|
||||
13273020
|
Waters Corporation
186003115
|
100 mm | 4.6 mm | 5 μm |
£1039.5
Each |
|
||||
15315255
|
Waters Corporation
186003116
|
150 mm | 4.6 mm | 5 μm |
£1108.5
Each |
|
||||
13223030
|
Waters Corporation
186003126
|
50 mm | 1 mm | 3.5 μm |
£999.0
Each |
|
||||
13233030
|
Waters Corporation
186003128
|
150 mm | 1 mm | 3.5 μm |
£1149.0
Each |
|
||||
13273030
|
Waters Corporation
186003132
|
100 mm | 3 mm | 5 μm |
£1039.5
Each |
|
||||
10537407
|
Waters Corporation
186003109
|
100 mm | 2.1 mm | 5 μm |
£1039.5
Each |
|
||||
11926938
|
Waters Corporation
186003110
|
150 mm | 2.1 mm | 5 μm |
£1108.5
Each |
|
||||
10034645
|
Waters Corporation
186003111
|
30 mm | 3 mm | 5 μm |
£942.0
Each |
|
||||
13243020
|
Waters Corporation
186003112
|
150 mm | 3 mm | 5 μm |
£1108.5
Each |
|
||||
10004853
|
Waters Corporation
186003127
|
100 mm | 1 mm | 5 μm |
£1074.0
Each |
|
||||
13243030
|
Waters Corporation
186003129
|
30 mm | 2.1 mm | 5 μm |
£942.0
Each |
|
||||
13263030
|
Waters Corporation
186003131
|
50 mm | 3 mm | 5 μm |
£964.5
Each |
|
||||
13283030
|
Waters Corporation
186003133
|
250 mm | 3 mm | 5 μm |
£1218.0
Each |
|
||||
13203040
|
Waters Corporation
186003135
|
30 mm | 4.6 mm | 5 μm |
£942.0
Each |
|
||||
13233270
|
Waters Corporation
186003943
|
250 mm | 4.6 mm | 3.5 μm |
£1252.5
Each |
|
Waters Corporation XBridge C18 Column
As a workhorse for HPLC method development, XBridge BEH C18 sorbent can be used over an entire range of mobile phase pH (1 to 12).
£895.5 - £1252.5
Specification
Max. Pressure | 6000 psi (415 Bar) |
---|---|
Particle Shape | Spherical |
Pore Size | 130 Å |
Quantity | 1/Ea. |
USP Type | L1 |
Specification
Max. Pressure | 6000 psi (415 Bar) |
---|---|
Particle Shape | Spherical |
Pore Size | 130 Å |
Quantity | 1/Ea. |
USP Type | L1 |
Endcapped | Yes |
Type | Column |