Kautex™ Screw Closures
Manufactured with polypropylene for excellent chemical resistance. Kautex™ Screw Closures are rugged caps designed for Kautex™ wide mouth HDPE bottles.
Additional Details:
Additional Details: Weight: 0.02700kg
Product Code. 10095851
Quantity Price
1 £ 2.6500000000000004 / Each
Estimated Shipment
14-07-2025
Add to basket
  • Description and Specification
  • Accessories and Spare Parts
Description and Specification
Specification
Color Blue
Includes Tamper Evident Ring and Conical Seal
Product Type Screw Cap
For Use With (Equipment) 1500/2500/4000 mL Bottle
Diameter (Metric) 80 mm
  • UN approval for solids when closures are used with associated bottles
  • Tamper evident ring
  • Conical seal provides leakproof closure
  • Available to fit bottles with capacities from 50 to 4000 mL
Accessories and Spare Parts