205.20 GBP valid until 2025-08-31
Use promo code "24157" to get your promotional price.
Use promo code "24157" to get your promotional price.
Closure Color | Clear |
---|---|
Closure Included | Yes |
Closure Material | Polystyrene |
Bottom Shape | Flat |
Capacity (Metric) | 0.57 mL (Well) |
Color | Clear |
Culture Area | 1.9 cm2 |
Material | Polystyrene |
Shape | Rectangular |
Sterility | Sterile |
Surface Treatment | CellBIND |
Type | Multi-well Cell Culture Plate |
Volume (Metric) Working | 0.38 to 0.57 mL |
Well Count | 24 |
Includes | Lid |
No. per Case | 50/Cs. |
No. per Pack | 5/Pk. |