Thermo Scientific Acros Ethanol, 95+%, Thermo Scientific Chemicals
Overview
302.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: 29
Additional Details:
Additional Details: CAS Number: 64-17-5 Weight: 8.00000kg Transport: UN number : 1170 Chem class : 3 Pack group : II
Quantity 10 L
Disclaimers: For Manufacturing and Laboratory Use. Not for beverage use
Product Code. 10329323
Quantity Price
1 £ 303.51 / 10 L
Estimated Shipment
29-05-2025
Add to basket
Sea freight only: For EU stock allow up to 15 days delivery.
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
Chemical Identifiers
CAS 64-17-5
Molecular Formula C2H6O
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym grain alcohol,algrain,ethyl hydrate,ethyl hydroxide,methylcarbinol,ethyl alcohol,alcohol,tecsol,anhydrol,alcohol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO
Specification
Chemical Name or Material Ethanol
Melting Point -117.0°C
CAS 64-17-5
CAS 67-56-1
Density 0.8000 g/mL
Boiling Point 78.0°C
Quantity 10 L
Flash Point 12°C
Packaging Plastic Drum
Refractive Index 1.3610 to 1.3630
Specific Gravity 0.8
Molecular Formula C2H6O
Linear Formula C2H5OH
Synonym grain alcohol,algrain,ethyl hydrate,ethyl hydroxide,methylcarbinol,ethyl alcohol,alcohol,tecsol,anhydrol,alcohol
Synonym grain alcohol,algrain,ethyl hydrate,ethyl hydroxide,methylcarbinol,ethyl alcohol,alcohol,tecsol,anhydrol,alcohol
Synonym grain alcohol,algrain,ethyl hydrate,ethyl hydroxide,methylcarbinol,ethyl alcohol,alcohol,tecsol,anhydrol,alcohol
Synonym grain alcohol,algrain,ethyl hydrate,ethyl hydroxide,methylcarbinol,ethyl alcohol,alcohol,tecsol,anhydrol,alcohol
Synonym grain alcohol,algrain,ethyl hydrate,ethyl hydroxide,methylcarbinol,ethyl alcohol,alcohol,tecsol,anhydrol,alcohol
Synonym grain alcohol,algrain,ethyl hydrate,ethyl hydroxide,methylcarbinol,ethyl alcohol,alcohol,tecsol,anhydrol,alcohol
Synonym grain alcohol,algrain,ethyl hydrate,ethyl hydroxide,methylcarbinol,ethyl alcohol,alcohol,tecsol,anhydrol,alcohol
Synonym grain alcohol,algrain,ethyl hydrate,ethyl hydroxide,methylcarbinol,ethyl alcohol,alcohol,tecsol,anhydrol,alcohol
Synonym grain alcohol,algrain,ethyl hydrate,ethyl hydroxide,methylcarbinol,ethyl alcohol,alcohol,tecsol,anhydrol,alcohol
Synonym grain alcohol,algrain,ethyl hydrate,ethyl hydroxide,methylcarbinol,ethyl alcohol,alcohol,tecsol,anhydrol,alcohol
Solubility Information Solubility aq. soln.: miscible. Other solubilities: miscible with many organic solvents
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.07
PubChem CID 702
ChEBI CHEBI:16236
Formula Weight 46.06
Grade Pure