Applied Biosystems™ SYBR™ Green Universal Master Mix
Everything needed for SYBR™ Green dye-based PCR amplification and detection in a convenient, single-tube format.
Overview
526.69 GBP valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Code: 50
Additional Details:
Additional Details: Weight: 0.02000kg
Disclaimers: For Research Use Only. Not for use in diagnostic procedures.
Product Code. 10418454
Quantity Price
1 £ 790.0 / 1 set
In stock, ready to ship: 51
Add to basket
For Research Use Only. All usage must comply with product instructions.
  • Description and Specification
  • Alternative Products
  • Special Offers
Description and Specification
Specification
Content And Storage The 2 x 5 mL vials contain a 2X mixture of SYBR Green 1 Dye, AmpliTaq Gold™ DNA Polymerase, dNTPs with dUTP, Passive Reference 1 (ROX), and optimized buffer components. Sufficient reagents provided for 400 reactions based on a 50 μl reaction volume.
Store at -20°C.
Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
Detection Method SYBR
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, QuantStudio™, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System
Label or Dye SYBR Green
Passive Reference Dye ROX (Pre-mixed)
Product Line SYBR™
Product Type Real Time PCR SYBR Master Mix
Quantity 2 x 5 mL
Sufficient For 400 Reactions of 50 μL
For Use With (Application) Gene Expression
Concentration 2X
No. of Reactions 400 Reactions
Sample Type DNA (Genomic), cDNA
Volume 2 x 5 mL
  • SYBR™ Green I dye detects double-stranded DNA, so specific probes are not required
  • AmpliTaq Gold™ DNA polymerase minimizes nonspecific product formation to achieve superior performance
  • dUTP significantly reduces carryover contamination when used in conjunction with uracil-DNA glycosylase
  • Proprietary buffer enhancements ensure performance and reliability
  • Premixed components stored at 2° to 8°C significantly reduce assay setup time
Maximum Flexibility and Convenience:
  • Applied Biosystems™ SYBR™ Green Universal Master Mix provides maximum flexibility at reduced cost because no target-specific TaqMan™ probes are required
  • SYBR™ Green I dye is a double-stranded DNA binding dye that detects any double-stranded DNA generated during PCR
  • The hot-start enzyme AmpliTaq Gold™ DNA Polymerase minimizes nonspecific product formation (including primer-dimers), yielding superior performance and sensitivity
  • Passive Internal Reference 1 is provided to normalize non-PCR—related fluorescence fluctuations
  • This minimizes well-to-well variability that can result from a variety of causes, such as pipetting error or sample evaporation
  • SYBR™ Green I dye is ideal for target identification (screening assays) or when a limited number of assays is needed.
  • Store at -20°C