Thermo Scientific™ TraceGOLD™ TG-WaxMS GC Columns
Analyze FAMEs with excellent thermal stability using this column.
Overview
369.42 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: 16
Additional Details:
Additional Details: Weight: 0.50000kg
Disclaimers: Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Code. 10419444
Quantity Price
1 £ 390.0 / Each
Estimated Shipment
23-06-2025
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
Specification
Max. Temperature 240/260°C
Diameter (Metric) 0.25 mm
Diameter (Metric) Inner 0.25 mm
Product Line TraceGOLD
Stationary Phase TraceGOLD™ TG-WAXMS
USP Type G14, G15, G16, G20, G39
For Use With (Application) Xylene Isomers Analysis,GC/MS for Flavor Compoundsand Essential Oils,GC/MS for Solvents,GC/MS for Xylene Isomers,GC/MS for EPA Method 603 for Acrolein/Acrylonitrile
For Use With (Application) Xylene Isomers Analysis,GC/MS for Flavor Compoundsand Essential Oils,GC/MS for Solvents,GC/MS for Xylene Isomers,GC/MS for EPA Method 603 for Acrolein/Acrylonitrile
For Use With (Application) Xylene Isomers Analysis,GC/MS for Flavor Compoundsand Essential Oils,GC/MS for Solvents,GC/MS for Xylene Isomers,GC/MS for EPA Method 603 for Acrolein/Acrylonitrile
For Use With (Application) Xylene Isomers Analysis,GC/MS for Flavor Compoundsand Essential Oils,GC/MS for Solvents,GC/MS for Xylene Isomers,GC/MS for EPA Method 603 for Acrolein/Acrylonitrile
For Use With (Application) Xylene Isomers Analysis,GC/MS for Flavor Compoundsand Essential Oils,GC/MS for Solvents,GC/MS for Xylene Isomers,GC/MS for EPA Method 603 for Acrolein/Acrylonitrile
Film Thickness 0.25 μm
Length (Metric) 15 m
Polarity Polar
Description 0.25 um Thickness; 0.25 mm ID; 15 m Length
Engineered for Superior Performance
  • Polar phase, polyethylene glycol
  • Polar-deactivated surface that tightly binds polymer for excellent thermal stability
  • Resistive to oxidative damage and damage from strongly acidic or basic volatiles better than silicone solid phases
  • Applications:
  • Fatty acid methyl esters (FAMEs)
  • Flavor compounds and essential oils
  • Solvents
  • Xylene isomers
  • EPA Method 603 for Acrolein/Acrylonitrile
A Leap Forward in Column Performance with Low Bleed and Superior Inertness
  • This low-bleed, stable column produces consistent results.
  • Low baseline noise enables improved limits of detection with enhanced resolution of low level analytes.
  • Minimizing column bleed extends column lifetime, leading to higher productivity.
  • Reducing column bleed increases reproducibility of the stationary phase over time, resulting in greater run-to-run and batch-to-batch reproducibility.
  • Superior inertness ensures excellent peak shape and sensitivity, especially for highly active or difficult-to-analyze compounds.
Recommended For:
  • Aldehydes
  • Essential oils
  • Solvents
  • Xylenes
  • Alternative Uses:
  • Acids
  • Alcohols
  • Alcohols in beverages
  • Alditol acetates (sugars)
  • Amines&endash;Aliphatic
  • Amines&endash;Aromatic
  • Herbicides
  • Ketones
  • Nitroaromatics
  • Phenols
  • Volatiles
Similar To:
  • DB-WAX
  • DB-WAXetr
  • HP-Wax
  • HP-Innowax
  • SUPELCOWAX™ 10
  • CP-Wax 52 CB
  • Stabilwax™
  • Rtx™-Wax
  • BP20
  • ZB-Wax
  • Optima™ Wax
  • AT™-Wax