926.93 GBP valid until 2025-06-30
Use promo code "25075" to get your promotional price.
Use promo code "25075" to get your promotional price.
Barcode | No Barcode |
---|---|
Certifications/Compliance | Certified RNase and DNase Free. ANSI Compliant. |
Format | Standard |
Lid | Without Lid |
Description | 384-well Polypropylene Sample Processing & Storage Plates |
Array | 16 x 24 |
Autoclavable | Autoclavable |
Packaging | 100 Cases |
Binding Type | Low binding PP |
Product Line | Nunc |
Color | White |
Product Type | 384-well plate |
For Use With (Equipment) | ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers |
Material | Polypropylene |
Volume (Metric) Well | 55 μL |
No. of Wells | 384 |
Volume (Metric) Working | 2 to 35 μL |
Sterility | Non-sterile |
Surface Treatment | Non-treated |
Well Shape | Square, Round Bottom (U-well) |
Dnase Free | Yes |
For Use With (Application) | Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation |
No. per Case | 100 |
No. per Pack | 25 |
Regulatory Status | For Research Use Only. Not for use in diagnostic procedures. |
RNase Free | Yes |
Fit for Purpose | Certified Quality |
Plate Size | ANSI/ SLAS Standard |