Use promo code "25339" to get your promotional price.
Thermo Scientific Alfa Aesar Ethanolamine, ACS, 99+%, Thermo Scientific Chemicals
Use promo code "25339" to get your promotional price.
Quantity | 500 mL |
---|
Chemical Identifiers
CAS | 141-43-5 |
---|---|
Molecular Formula | C2H7NO |
Molecular Weight (g/mol) | 61.08 |
MDL Number | MFCD00008183 |
InChI Key | HZAXFHJVJLSVMW-UHFFFAOYSA-N |
Synonym | ethanolamine,monoethanolamine,aminoethanol,2-hydroxyethylamine,colamine,glycinol,olamine,2-amino-1-ethanol,ethanol, 2-amino,ethylolamine |
PubChem CID | 700 |
ChEBI | CHEBI:16000 |
SMILES | NCCO |
Specification
Chemical Name or Material | Ethanolamine |
---|---|
Name Note | For analysis ACS |
CAS | 141-43-5 |
Melting Point | 10°C |
Density | 1.012 g/mL |
Boiling Point | 169°C to 170°C |
Flash Point | 93°C (199°F) |
Assay Percent Range | >99+% |
Molecular Formula | C2H7NO |
Refractive Index | 1.454 |
MDL Number | MFCD00008183 |
Quantity | 500 mL |
UN Number | UN2491 |
Beilstein | 505944 |
Sensitivity | Air sensitive; Hygroscopic |
Merck Index | 14,3727 |
Synonym | ethanolamine,monoethanolamine,aminoethanol,2-hydroxyethylamine,colamine,glycinol,olamine,2-amino-1-ethanol,ethanol, 2-amino,ethylolamine |
Synonym | ethanolamine,monoethanolamine,aminoethanol,2-hydroxyethylamine,colamine,glycinol,olamine,2-amino-1-ethanol,ethanol, 2-amino,ethylolamine |
Synonym | ethanolamine,monoethanolamine,aminoethanol,2-hydroxyethylamine,colamine,glycinol,olamine,2-amino-1-ethanol,ethanol, 2-amino,ethylolamine |
Synonym | ethanolamine,monoethanolamine,aminoethanol,2-hydroxyethylamine,colamine,glycinol,olamine,2-amino-1-ethanol,ethanol, 2-amino,ethylolamine |
Synonym | ethanolamine,monoethanolamine,aminoethanol,2-hydroxyethylamine,colamine,glycinol,olamine,2-amino-1-ethanol,ethanol, 2-amino,ethylolamine |
Synonym | ethanolamine,monoethanolamine,aminoethanol,2-hydroxyethylamine,colamine,glycinol,olamine,2-amino-1-ethanol,ethanol, 2-amino,ethylolamine |
Synonym | ethanolamine,monoethanolamine,aminoethanol,2-hydroxyethylamine,colamine,glycinol,olamine,2-amino-1-ethanol,ethanol, 2-amino,ethylolamine |
Synonym | ethanolamine,monoethanolamine,aminoethanol,2-hydroxyethylamine,colamine,glycinol,olamine,2-amino-1-ethanol,ethanol, 2-amino,ethylolamine |
Synonym | ethanolamine,monoethanolamine,aminoethanol,2-hydroxyethylamine,colamine,glycinol,olamine,2-amino-1-ethanol,ethanol, 2-amino,ethylolamine |
Synonym | ethanolamine,monoethanolamine,aminoethanol,2-hydroxyethylamine,colamine,glycinol,olamine,2-amino-1-ethanol,ethanol, 2-amino,ethylolamine |
Solubility Information | Miscible in water. |
InChI Key | HZAXFHJVJLSVMW-UHFFFAOYSA-N |
SMILES | NCCO |
IUPAC Name | 2-aminoethan-1-ol |
Molecular Weight (g/mol) | 61.08 |
PubChem CID | 700 |
ChEBI | CHEBI:16000 |
Formula Weight | 61.08 |
Percent Purity | ≥99% |
Grade | ACS |
Physical Form | Liquid |
Technical documents
[Ethanolamine, ACS]
[Danger]
[{code=ACUTE TOX. 4, sequence=1, description=Acute toxicity Category 4}, {code=AQUA CHRONIC 3, sequence=2, description=LONG-TERM AQUATIC HAZARD Chronic 3}, {code=EYE DAM. 1, sequence=3, description=Serious eye damage/eye irritation Category 1}, {code=SKIN CORR. 1B, sequence=4, description=Skin corrosion/irritation Category 1 B}, {code=STOT SE 3, sequence=5, description=Specific target organ toxicity Category 3}]
[{code=H302, sequence=1, description=Harmful if swallowed.}, {code=H312, sequence=2, description=Harmful in contact with skin.}, {code=H314, sequence=3, description=Causes severe skin burns and eye damage.}, {code=H318, sequence=4, description=Causes serious eye damage.}, {code=H332, sequence=5, description=Harmful if inhaled.}, {code=H335, sequence=6, description=May cause respiratory irritation.}, {code=H412, sequence=7, description=Harmful to aquatic life with long lasting effects.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=52, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P303+P361+P353, sequence=53, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P304+P340, sequence=54, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=55, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=56, description=Immediately call a POISON CENTER/doctor/.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : Ethanolamine}]
[{code=GHS05, sequence=1, description=CORROSION}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]
Product Identifier
- Ethanolamine, ACS
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H302- Harmful if swallowed.
- H312- Harmful in contact with skin.
- H314- Causes severe skin burns and eye damage.
- H318- Causes serious eye damage.
- H332- Harmful if inhaled.
- H335- May cause respiratory irritation.
- H412- Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.
Supplemental Information
- MIXTURE LIST- Contains : Ethanolamine