Code:
04
VistaLab™ Technologies Fixed Volume Ovation BioNatural Pipettes
Attain accurate pipetting with this single stroke pipette. Vistalab™ Fixed Volume Ovation BioNatural Pipettes are excellent for pipetting exact volume of liquids without deviations.
Additional Details:
Weight:
0.01000kg
Specification
Description | Single stroke |
---|---|
Color | Purple |
Material | Polycarbonate ABS |
Model | F1 |
Number of Channels | 1 |
Type | Fixed Volume Ovation Bionatural Pipette |
Volume (Metric) | 200 μL |
Precision | 0.5% |
Quantity | 1 ea. |
Warranty | 2 Year Limited Warranty |
Length (English) | 9 in. |
Width (English) | 7 in. |
Height (English) | 3 in. |