Ansell HyFlex™ 11-738 Intercept™ Cut-Resistant Gloves with Targeted Reinforcement
Fortix™ Technology increases protection with additional durability in the thumb crotch region, and extends the glove's useful life.
Overview
176.04 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Additional Details:
Additional Details: Weight: 9.07000kg
Product Code. 16009661
Quantity Price
1 £ 246.0 / 12 pairs
Estimated Shipment
21-10-2025
Specification
ANSI Abrasion Rating ANSI 5
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A4
Hand Specific
Latex Free Yes
Material Intercept Technology Yarn
Product Type Cut-resistant Gloves
Size 7
EN388 Standard 4X42C
For Use With (Application) Automotive Assembly, Machinery, Equipment Operation, and Metal Fabrication
Glove Cuff Style Knit
Series 11-738
Yarn Gauge 13
  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Fortix™ Technology extends product's useful life
  • Increased protection in the thumb crotch region
  • Water-based polyurethane enhances comfort and dexterity
  • Oeko-Tek™ 100 Certified
  • Solvent-free
Alternative Products