Waters Corporation XSelect CSH Phenyl-Hexyl Column
XSelect CSH Phenyl-Hexyl columns provide you with alternative analyte selectivity compared to traditional reversed-phase columns and are a valuable tool for method development.
Additional Details:
Weight:
0.10200kg
Specification
Diameter (Metric) | 3 mm |
---|---|
Length (Metric) | 30 mm |
Max. Pressure | 6000 psi (415 Bar) |
Particle Shape | Spherical |
Particle Size | 5 μm |
Pore Size | 130 Å |
Quantity | 1/Ea. |
USP Type | L11 |
Endcapped | Yes |
Type | Column |