Greiner Bio-One 96-well Polystyrene Round Bottom Strip Plates, 2 x 8 Strips
Treated specifically to increase protein binding. Greiner Bio-One 96 Well Polystyrene Round Bottom Strip Plates, 2 x 8 Strips are suited for agglutination tests.
Additional Details:
Weight:
0.80000kg
Specification
Format | Strip Plate |
---|---|
Binding Type | High Binding |
Color | White |
Material | Polystyrene |
No. of Wells | 6 |
Volume (Metric) Working | 50 to 280 μL |
Sterility | Non-sterile |
Surface Treatment | Microlon 600 |
Total Volume (Metric) Well | 312 μL |
Well Shape | Round Bottom (U-well) |
Dnase Free | Yes |
No. per Case | 100 |
No. per Pack | 5 |
RNase Free | Yes |
Temperature Range (Metric) | -20°C to +60°C |
Well Color | Clear |