Code:
67
Corning™ BioCoat™ Collagen I, Rat Tail Culture Dish
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Additional Details:
Weight:
4.76270kg
Specification
Closure Color | Clear |
---|---|
Closure Included | Closure Included |
Closure Material | Polystyrene |
Bottom Shape | Round |
Color | Clear |
Culture Area | 58.1 sq. cm |
Material | Polystyrene |
Shape | Round |
Surface Treatment | Collagen Type I |
Type | Cell Culture Dish |
Includes | Lid |
Quantity | 10/cs. |
Diameter (Metric) | 100 mm |