Proteintech™ HumanKine™ Recombinant Human TGF Beta 3 Protein
HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells
Additional Details:
Weight:
0.00010kg
Specification
| Gene ID (Entrez) | 7043 |
|---|---|
| Molecular Weight (g/mol) | 25 kDa, homodimer, non-glycosylated |
| Quantity | 1000 μg |
| Species | Human |
| Reconstitution | Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 4 mM HCl containing 0.1% endotoxin-free recombinant human serum albumin (HSA). |
| Content And Storage | Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. |
| Activity | Typically ≤ 0.5 ng/mL EC50 |
| Expression System | Human embryonic kidney 293 (HEK293) human cell expression system |
| Form | Lyophilized |
| Purity or Quality Grade | > 95% |