Proteintech™ HumanKine™ Recombinant Human TGF Beta 3 Protein

HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells

£70.0 - £2845.0

Specification

Gene ID (Entrez) 7043
Molecular Weight (g/mol) 25 kDa, homodimer, non-glycosylated
Species Human
Reconstitution Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 4 mM HCl containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Content And Storage Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
View More Specs


Specification

Gene ID (Entrez) 7043
Molecular Weight (g/mol) 25 kDa, homodimer, non-glycosylated
Species Human
Reconstitution Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 4 mM HCl containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Content And Storage Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Activity Typically ≤ 0.5 ng/mL EC50
Expression System Human embryonic kidney 293 (HEK293) human cell expression system
Form Lyophilized
Purity or Quality Grade > 95%

  • Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity

Product Code Brand Quantity Price ??product.searchResults.label.quantityAvail??
Quantity
16861306
Proteintech™
HZ-1090-5UG
5 μg £70.0
5µg
Add to basket
16851306
Proteintech™
HZ-1090-100UG
100 μg £570.0
100µg
Add to basket
16841306
Proteintech™
HZ-1090-1000UG
1000 μg £2845.0
1000µg
Add to basket