281.65 GBP valid until 2025-04-30
Use promo code "24259" to get your promotional price.
Use promo code "24259" to get your promotional price.
Product Type | Biohazard Bag |
---|---|
Autoclavable | Autoclavable |
Capacity (English) | 20 gal. |
Capacity (Metric) | 170 L |
Certifications/Compliance | ASTM D1709-98 |
Closure Type | Zip Tie |
Color | Translucent |
Disposable | Single-use |
For Use With (Application) | Disposal of used pipettes, petri dishes, culture plates, membrane filters and more |
Includes | Non-printed without Sterilization Patch |
Material | Polypropylene |
Sterility | Non-sterile |
Thickness (Metric) | 1.5 mm |
Thickness (English) | 1.5 mil |
Dimensions (L x W) | 36 x 24 in. (91 x 61 cm) |
Length (English) | 36 in. |
Length (Metric) | 91 cm |
Width (English) | 24 in. |
Width (Metric) | 61 cm |